missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromotriphenylamine, 98%

Catalog No. AAH6466603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6466603 1 g
AAH6466606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6466603 Supplier Thermo Scientific Chemicals Supplier No. H6466603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 81090-53-1 | C18H13Br2N | 403.117 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 81090-53-1
Molecular Formula C18H13Br2N
Molecular Weight (g/mol) 403.117
MDL Number MFCD00060104
InChI Key KIGVOJUDEQXKII-UHFFFAOYSA-N
Synonym 4,4'-dibromotriphenylamine, 4-bromo-n-4-bromophenyl-n-phenylaniline, n,n-bis 4-bromophenyl aniline, 4,4-dibromotriphenylamine, 4-bromo-n-4-bromophenyl-n-phenylbenzenamine, dibromotriphenylamine, 4,4'-dibromotriphenylamine;, 4,4'-dibromo-triphenylamine, bis 4-bromophenyl phenylamine, bis 4-bromophenyl phenylamine;
PubChem CID 13594707
IUPAC Name 4-bromo-N-(4-bromophenyl)-N-phenylaniline
SMILES C1=CC=C(C=C1)N(C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br

Specifications

Melting Point 85°C
Color White
Flash Point >110°C (230°F)
Quantity 1 g
Beilstein 7094354
Formula Weight 403.12
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4,4'-Dibromotriphenylamine
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only