missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromooctafluorobiphenyl, 99%

Catalog No. p-7028016
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2512606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2512606 Supplier Thermo Scientific Chemicals Supplier No. B2512606
Only null left

CAS: 10386-84-2 | C12Br2F8 | 455.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10386-84-2
Molecular Formula C12Br2F8
Molecular Weight (g/mol) 455.93
MDL Number MFCD00000310
InChI Key YXLMNFVUNLCJJY-UHFFFAOYSA-N
PubChem CID 82600
IUPAC Name 1-bromo-4-(4-bromo-2,3,5,6-tetrafluorophenyl)-2,3,5,6-tetrafluorobenzene
SMILES FC1=C(F)C(=C(F)C(F)=C1Br)C1=C(F)C(F)=C(Br)C(F)=C1F

Specifications

Melting Point 113°C to 115°C
Quantity 5 g
UN Number UN3152
Synonym 4,4'-dibromooctafluorobiphenyl, 4,4'-dibromo-2,2',3,3',5,5',6,6'-octafluorobiphenyl, octafluoro-4,4'-dibromobiphenyl, 4,4'-dibromo-2,2',3,3',5,5',6,6'-octafluoro-1,1'-biphenyl, 1,1'-biphenyl, 4,4'-dibromo-2,2',3,3',5,5',6,6'-octafluoro, 1,1'-biphenyl,4,4'-dibromo-2,2',3,3',5,5',6,6'-octafluoro, 1-bromo-4-4-bromo-2,3,5,6-tetrafluorophenyl-2,3,5,6-tetrafluorobenzene, f-pbb, 4,4/'-dibromooctafluorobiphenyl, 4,4\\'-dibromooctafluorobiphenyl
Formula Weight 455.94
Percent Purity 99%
Chemical Name or Material 4,4'-Dibromooctafluorobiphenyl
DOTInformation Transport Hazard Class: 9; Packing Group: II; Proper Shipping Name: POLYHALOGENATED BIPHENYLS, SOLID
EINECSNumber 233-847-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only