missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromobiphenyl, 98+%

Catalog No. p-7021697
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1413918 50 g
AAA1413930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1413918 Supplier Thermo Scientific Chemicals Supplier No. A1413918

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 92-86-4 | C12H8Br2 | 312.00 g/mol

4,4'-Dibromobiphenyl is commonly used as fire retardants and plasticizers. It is also have application in gas chromatography and liquid chromatography analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dibromobiphenyl is commonly used as fire retardants and plasticizers. It is also have application in gas chromatography and liquid chromatography analysis.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 92-86-4
Molecular Formula C12H8Br2
Molecular Weight (g/mol) 312.00
MDL Number MFCD00000101
InChI Key HQJQYILBCQPYBI-UHFFFAOYSA-N
Synonym 4,4'-dibromobiphenyl, 1,1'-biphenyl, 4,4'-dibromo, p,p'-dibromobiphenyl, 4,4'-dibromodiphenyl, 1-bromo-4-4-bromophenyl benzene, 4,4'-dibromo-biphenyl, unii-pou26v2xt2, 4',4-dibromobiphenyl, biphenyl, 4,4'-dibromo, pou26v2xt2
PubChem CID 7110
IUPAC Name 1-bromo-4-(4-bromophenyl)benzene
SMILES BrC1=CC=C(C=C1)C1=CC=C(Br)C=C1

Specifications

Melting Point 164°C to 169°C
Boiling Point 355°C to 360°C
Quantity 50 g
UN Number UN3152
Beilstein 2044701
Solubility Information Insoluble in water.
Formula Weight 312.02
Percent Purity ≥98%
Chemical Name or Material 4,4'-Dibromobiphenyl
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: II; Proper Shipping Name: POLYHALOGENATED BIPHENYLS, SOLID
EINECSNumber 202-198-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only