missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dibromo-trans-stilbene 98.0+%, TCI America™
$33.56 - $266.56
Chemical Identifiers
| CAS | 18869-30-2 |
|---|---|
| Molecular Formula | C14H10Br2 |
| Molecular Weight (g/mol) | 338.04 |
| MDL Number | MFCD00448009 |
| InChI Key | JEHMPNUQSJNJDL-UHFFFAOYSA-N |
| PubChem CID | 7020566 |
| IUPAC Name | 1-bromo-4-[2-(4-bromophenyl)ethenyl]benzene |
| SMILES | BrC1=CC=C(C=CC2=CC=C(Br)C=C2)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D3043250MG
|
TCI America
D3043250MG |
250 mg |
Each for $33.56
|
|
|||||
|
D30431G
|
TCI America
D30431G |
1 g |
Each for $89.48
|
|
|||||
|
D30435G
|
TCI America
D30435G |
5 g |
Each for $266.56
|
|
|||||
Chemical Identifiers
| 18869-30-2 | |
| 338.04 | |
| JEHMPNUQSJNJDL-UHFFFAOYSA-N | |
| 1-bromo-4-[2-(4-bromophenyl)ethenyl]benzene |
| C14H10Br2 | |
| MFCD00448009 | |
| 7020566 | |
| BrC1=CC=C(C=CC2=CC=C(Br)C=C2)C=C1 |
Specifications
| 18869-30-2 | |
| White-Yellow | |
| MFCD00448009 | |
| JEHMPNUQSJNJDL-UHFFFAOYSA-N | |
| 1-bromo-4-[2-(4-bromophenyl)ethenyl]benzene | |
| 7020566 | |
| ≥98.0% (GC) | |
| 4,4′-Dibromo-trans-stilbene |
| 212°C | |
| C14H10Br2 | |
| 250 mg | |
| BrC1=CC=C(C=CC2=CC=C(Br)C=C2)C=C1 | |
| 338.04 | |
| 338.04 | |
| Crystalline Powder |
Safety and Handling
TSCA : No