missing translation for 'onlineSavingsMsg'
Learn More

4,4″-Dibromo-p-terphenyl 95.0+%, TCI America™

Catalog No. D33725G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33725G 5 g
D33721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33725G Supplier TCI America Supplier No. D33725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17788-94-2
Molecular Formula C18H12Br2
Molecular Weight (g/mol) 388.10
MDL Number MFCD00272889
InChI Key VAIPJQIPFPRJKJ-UHFFFAOYSA-N
Synonym 4,4-dibromo-p-terphenyl, 4,4-dibromo-1,1':4',1-terphenyl, 1,1':4',1-terphenyl, 4,4-dibromo, acmc-1c6h6, 1,4-bis 4-bromophenyl benzene, 4-bromo-4'-4-bromophenyl-1,1'-biphenyl, 1∼4∼,3∼4∼-dibromo-1∼1∼,2∼1∼:2∼4∼,3∼1∼-terphenyl, 4,4 inverted exclamation marka inverted exclamation marka-dibromo-p-terphenyl, 4,4 inverted exclamation marka inverted exclamation marka-dibromo-1,1 inverted exclamation marka:4 inverted exclamation marka,1 inverted exclamation marka inverted exclamation marka-terphenyl
PubChem CID 12455998
IUPAC Name 4-bromo-4'-(4-bromophenyl)-1,1'-biphenyl
SMILES BrC1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(Br)C=C1

Specifications

Melting Point 315°C
Quantity 5 g
UN Number 3152
Formula Weight 388.10
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′′-Dibromo-p-terphenyl