missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromo-2-nitrobiphenyl 98.0+%, TCI America™

Catalog No. D51891G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51891G 1 g
D51895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51891G Supplier TCI America Supplier No. D51891G

Chemical Identifiers

CAS 439797-69-0
Molecular Formula C12H7Br2NO2
Molecular Weight (g/mol) 357.001
MDL Number MFCD05854318
InChI Key FMSJGXRUJCWSJL-UHFFFAOYSA-N
PubChem CID 1519133
IUPAC Name 4-bromo-1-(4-bromophenyl)-2-nitrobenzene
SMILES C1=CC(=CC=C1C2=C(C=C(C=C2)Br)[N+](=O)[O-])Br

Specifications

Melting Point 127°C
Color Yellow
Quantity 1 g
Formula Weight 357.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dibromo-2-nitrobiphenyl