missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromo-2,2'-bithiophene 98.0+%, TCI America™

Catalog No. D41811G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41811G 1 g
D41815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41811G Supplier TCI America Supplier No. D41811G

Chemical Identifiers

CAS 51285-60-0
Molecular Formula C8H4Br2S2
Molecular Weight (g/mol) 324.048
MDL Number MFCD00508561
InChI Key KITUXFRDWJKACE-UHFFFAOYSA-N
PubChem CID 716032
IUPAC Name 4-bromo-2-(4-bromothiophen-2-yl)thiophene
SMILES C1=C(SC=C1Br)C2=CC(=CS2)Br

Specifications

Melting Point 134°C
Color Yellow
Quantity 1 g
Formula Weight 324.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dibromo-2,2′-bithiophene