accessibility menu, dialog, popup

UserName

CAS RN 51285-60-0

IUPAC Name: 4,4'-dibromo-2,2'-bithiophene
Synonyms: 4,4'-dibromo-2,2'-bithiophene
Molecular Weight (g/mol): 324.05
Molecular Formula: C8H4Br2S2
InChi Key: KITUXFRDWJKACE-UHFFFAOYSA-N
SMILES: BrC1=CSC(=C1)C1=CC(Br)=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results