missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromo-2,2'-bipyridyl 97.0+%, TCI America™

Catalog No. D39195G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39195G 5 g
D39191G 1 g
D391925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D39195G Supplier TCI America Supplier No. D39195G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18511-71-2 | C10H6Br2N2 | 313.98 g/mol

Chemical Identifiers

CAS 18511-71-2
Molecular Formula C10H6Br2N2
Molecular Weight (g/mol) 313.98
MDL Number MFCD00667763
InChI Key KIIHBDSNVJRWFY-UHFFFAOYSA-N
Synonym 4,4'-dibromo-2,2'-bipyridine, 4-bromo-2-4-bromopyridin-2-yl pyridine, 4,4'-dibromo-2,2'-bipyridyl, 2,2'-bipyridine, 4,4'-dibromo, bpbb01, acmc-1bo83, 4,4'-dibromo-2,2-bipyridine, 4,4'-bromo-2,2'-bipyridine, 4,4'-dibromo-2,2'-dipyridine, 4,4'-dibromine-2,2'-bipyridine
PubChem CID 15569987
IUPAC Name 4-bromo-2-(4-bromopyridin-2-yl)pyridine
SMILES C1=CN=C(C=C1Br)C2=NC=CC(=C2)Br

Specifications

Melting Point 140°C
Color White-Yellow
Quantity 5 g
Formula Weight 313.98
Percent Purity ≥97.0% (GC,T)
Grade Reagent
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dibromo-2,2′-bipyridyl