accessibility menu, dialog, popup

UserName

NBrNBr

CAS RN 18511-71-2

IUPAC Name: 4,4'-dibromo-2,2'-bipyridine
Synonyms: 4,4'-dibromo-2,2'-bipyridine
Molecular Weight (g/mol): 313.98
Molecular Formula: C10H6Br2N2
InChi Key: KIIHBDSNVJRWFY-UHFFFAOYSA-N
SMILES: BrC1=CC(=NC=C1)C1=NC=CC(Br)=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)