missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromo-1,1'-spirobiindane-7,7'-diol 98.0+%, TCI America™

Catalog No. D43325G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43325G 5 g
D43321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43325G Supplier TCI America Supplier No. D43325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 681481-91-4
Molecular Formula C17H14Br2O2
Molecular Weight (g/mol) 410.105
MDL Number MFCD16660947
InChI Key OZVMVFKUNSNXQC-UHFFFAOYSA-N
PubChem CID 11235237
IUPAC Name 7,7'-dibromo-3,3'-spirobi[1,2-dihydroindene]-4,4'-diol
SMILES C1CC2(CCC3=C(C=CC(=C32)O)Br)C4=C(C=CC(=C41)Br)O

Specifications

Melting Point 148°C
Quantity 5 g
Synonym 4,4′-Dibromo-7,7′-dihydroxy-1,1′-spirobiindane
Formula Weight 410.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dibromo-1,1′-spirobiindane-7,7′-diol