missing translation for 'onlineSavingsMsg'
Learn More

4,4″-Dibromo-1,1':3',1″-terphenyl 98.0+%, TCI America™

Supplier:  TCI America D50231G

 View more versions of this product

Catalog No. D50231G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4,4′′-Dibromo-1,1′:3′,1′′-terphenyl
158°C
C18H12Br2
1 g
4,4′′-Dibromo-m-terphenyl
C1=CC(=CC(=C1)C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br
388.102
388.10
Crystalline Powder
83909-22-2
White
MFCD28138091
3152
XYRNOZKOAZFOOG-UHFFFAOYSA-N
1,3-bis(4-bromophenyl)benzene
10667976
≥98.0% (GC)

Chemical Identifiers

83909-22-2
388.102
XYRNOZKOAZFOOG-UHFFFAOYSA-N
10667976
C1=CC(=CC(=C1)C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br
C18H12Br2
MFCD28138091
4,4′′-Dibromo-m-terphenyl
1,3-bis(4-bromophenyl)benzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions