missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diaminostilbene Dihydrochloride 98.0+%, TCI America™

Catalog No. D01201G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D01201G 1 g
D01205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D01201G Supplier TCI America Supplier No. D01201G
Only null left

Chemical Identifiers

CAS 54760-75-7
Molecular Formula C14H16Cl2N2
Molecular Weight (g/mol) 283.196
MDL Number MFCD00012984
InChI Key QHDAFTKIEDDTPV-SEPHDYHBSA-N
PubChem CID 11954176
IUPAC Name 4-[(E)-2-(4-aminophenyl)ethenyl]aniline;dihydrochloride
SMILES C1=CC(=CC=C1C=CC2=CC=C(C=C2)N)N.Cl.Cl

Specifications

Quantity 1 g
Formula Weight 283.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Diaminostilbene Dihydrochloride