missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%

Catalog No. AC406411000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC406411000 100 g Glass bottle
AC406415000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC406411000 Supplier Thermo Scientific Chemicals Supplier No. 406411000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 81-11-8 | C14H14N2O6S2 | 370.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 81-11-8
Molecular Formula C14H14N2O6S2
Molecular Weight (g/mol) 370.4
MDL Number MFCD00024946
InChI Key REJHVSOVQBJEBF-OWOJBTEDSA-N
Synonym amsonic acid, flavonic acid, tinopal bhs, amsonsaeure, dasd, 4,4'-diaminostilbene-2,2'-disulfonic acid, unii-h68088wvjl, 2,2'-disulfo-4,4'-stilbenediamine, diaminostilbenedisulfonic acid, 4,4'-diaminostilbene-2,2'-disulphonic acid
PubChem CID 5284378
IUPAC Name 5-amino-2-[(E)-2-(4-amino-2-sulfophenyl)ethenyl]benzenesulfonic acid
SMILES C1=CC(=C(C=C1N)S(=O)(=O)O)C=CC2=C(C=C(C=C2)N)S(=O)(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point >300°C
Color Brown-Yellow to Cream
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 100 g
Beilstein 14,I,744
Merck Index 15, 590
Solubility Information Solubility in water: very slightly soluble. Other solubilities: soluble in ethanol,diethyl ether
Formula Weight 370.4
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4, 4'-Diaminostilbene-2, 2'-disulfonic acid, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-325-2
RTECSNumber WJ6603000
TSCA TSCA

RUO – Research Use Only