missing translation for 'onlineSavingsMsg'
Learn More

4,4-Diaminodiphenyl sulfide, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020353
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020353 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020353 Supplier Merck Emd Millipore Supplier No. 32955250MG
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 139-65-1
Physical Form Neat
Linear Formula S(C6H4 NH2)2
MDL Number MFCD00025342
Melting Point 105°C to 107°C
Synonym 4,4 ′-Thiodianiline
Type Analytical Standard
Quantity 250 mg
Formula Weight 216.30
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less