missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diaminobenzanilide 98.0+%, TCI America™

Catalog No. D2005500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2005500G 500 g
D200525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2005500G Supplier TCI America Supplier No. D2005500G

Chemical Identifiers

CAS 785-30-8
Molecular Formula C13H13N3O
Molecular Weight (g/mol) 227.27
MDL Number MFCD00025361
InChI Key XPAQFJJCWGSXGJ-UHFFFAOYSA-N
Synonym 4,4'-diaminobenzanilide, 4-amino-n-4-aminophenyl benzamide, benzamide, 4-amino-n-4-aminophenyl, 4-aminobenzoyl-4'-aminoanilide, p,p'-diaminobenzanilide, 4,4'-diamino benzanilide, unii-00x4jm89uh, benzanilide, 4,4'-diamino, 4-aminophenyl-n-4-aminophenyl carboxamide, ccris 8916
PubChem CID 69917
IUPAC Name 4-amino-N-(4-aminophenyl)benzamide
SMILES NC1=CC=C(NC(=O)C2=CC=C(N)C=C2)C=C1

Specifications

Quantity 500 g
Formula Weight 227.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Diaminobenzanilide
EINECSNumber (3)-1491
TSCA Yes