missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diacetylbiphenyl, 98%

CAS: 787-69-9 | C16H14O2 | 238.286 g/mol

$159.04 - $778.49

Chemical Identifiers

CAS 787-69-9
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.286
MDL Number MFCD00017248
InChI Key YSTSBXDVNKYPTR-UHFFFAOYSA-N
Synonym 4,4'-diacetylbiphenyl, 4,4'-diacetyl biphenyl, 1-4-4-acetylphenyl phenyl ethanone, 4,4-diacetylbiphenyl, 1,1'-1,1'-biphenyl-4,4'-diyl diethanone, 1-4'-acetyl 1,1'-biphenyl-4-yl ethanone, ethanone, 1,1'-1,1'-biphenyl-4,4'-diylbis, 1-acetyl-4-4-acetylphenyl benzene, 1-4'-acetyl-1,1'-biphenyl-4-yl ethanone, diacetyl biphenyl
PubChem CID 301558
IUPAC Name 1-[4-(4-acetylphenyl)phenyl]ethanone
SMILES CC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1591106
View Documents
Thermo Scientific Chemicals
A1591106
5 g
Each for $159.04
Only null left
 
AAA1591114
View Documents
Thermo Scientific Chemicals
A1591114
25 g
Each for $778.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

787-69-9
238.286
YSTSBXDVNKYPTR-UHFFFAOYSA-N
301558
CC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C
C16H14O2
MFCD00017248
4,4'-diacetylbiphenyl, 4,4'-diacetyl biphenyl, 1-4-4-acetylphenyl phenyl ethanone, 4,4-diacetylbiphenyl, 1,1'-1,1'-biphenyl-4,4'-diyl diethanone, 1-4'-acetyl 1,1'-biphenyl-4-yl ethanone, ethanone, 1,1'-1,1'-biphenyl-4,4'-diylbis, 1-acetyl-4-4-acetylphenyl benzene, 1-4'-acetyl-1,1'-biphenyl-4-yl ethanone, diacetyl biphenyl
1-[4-(4-acetylphenyl)phenyl]ethanone

Specifications

787-69-9
C16H14O2
5 g
YSTSBXDVNKYPTR-UHFFFAOYSA-N
1-[4-(4-acetylphenyl)phenyl]ethanone
301558
98%
190°C to 193°C
MFCD00017248
4,4'-diacetylbiphenyl, 4,4'-diacetyl biphenyl, 1-4-4-acetylphenyl phenyl ethanone, 4,4-diacetylbiphenyl, 1,1'-1,1'-biphenyl-4,4'-diyl diethanone, 1-4'-acetyl 1,1'-biphenyl-4-yl ethanone, ethanone, 1,1'-1,1'-biphenyl-4,4'-diylbis, 1-acetyl-4-4-acetylphenyl benzene, 1-4'-acetyl-1,1'-biphenyl-4-yl ethanone, diacetyl biphenyl
CC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C
238.286
238.29
4,4'-Diacetylbiphenyl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 9; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only