missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diacetylbiphenyl, 98%

CAS: 787-69-9 | C16H14O2 | 238.286 g/mol

Supplier:  Thermo Scientific Chemicals A1591114

 View more versions of this product

Catalog No. AAA1591114

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4,4'-Diacetylbiphenyl
190°C to 193°C
MFCD00017248
4,4'-diacetylbiphenyl, 4,4'-diacetyl biphenyl, 1-4-4-acetylphenyl phenyl ethanone, 4,4-diacetylbiphenyl, 1,1'-1,1'-biphenyl-4,4'-diyl diethanone, 1-4'-acetyl 1,1'-biphenyl-4-yl ethanone, ethanone, 1,1'-1,1'-biphenyl-4,4'-diylbis, 1-acetyl-4-4-acetylphenyl benzene, 1-4'-acetyl-1,1'-biphenyl-4-yl ethanone, diacetyl biphenyl
CC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C
238.286
238.29
787-69-9
C16H14O2
25 g
YSTSBXDVNKYPTR-UHFFFAOYSA-N
1-[4-(4-acetylphenyl)phenyl]ethanone
301558
98%

Chemical Identifiers

787-69-9
238.286
YSTSBXDVNKYPTR-UHFFFAOYSA-N
301558
CC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C
C16H14O2
MFCD00017248
4,4'-diacetylbiphenyl, 4,4'-diacetyl biphenyl, 1-4-4-acetylphenyl phenyl ethanone, 4,4-diacetylbiphenyl, 1,1'-1,1'-biphenyl-4,4'-diyl diethanone, 1-4'-acetyl 1,1'-biphenyl-4-yl ethanone, ethanone, 1,1'-1,1'-biphenyl-4,4'-diylbis, 1-acetyl-4-4-acetylphenyl benzene, 1-4'-acetyl-1,1'-biphenyl-4-yl ethanone, diacetyl biphenyl
1-[4-(4-acetylphenyl)phenyl]ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 9; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only