missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Di-tert-butylbiphenyl, 99%

CAS: 1625-91-8 | C20H26 | 266.428 g/mol

$1023.88 - $1023.88

Chemical Identifiers

CAS 1625-91-8
Molecular Formula C20H26
Molecular Weight (g/mol) 266.428
MDL Number MFCD00008834
InChI Key CDKCEZNPAYWORX-UHFFFAOYSA-N
Synonym 4,4'-di-tert-butylbiphenyl, 4,4'-di-tert-butyl-1,1'-biphenyl, 4,4'-di-t-butylbiphenyl, 1-tert-butyl-4-4-tert-butylphenyl benzene, 1,1'-biphenyl, 4,4'-bis 1,1-dimethylethyl, 1-tert-butyl-4-4-tert-butyl phenyl benzene, di-t-butylbiphenyl, pubchem9048, acmc-209st2, 4,4'-di-tertbutylbiphenyl
PubChem CID 74195
IUPAC Name 1-tert-butyl-4-(4-tert-butylphenyl)benzene
SMILES CC(C)(C)C1=CC=C(C=C1)C2=CC=C(C=C2)C(C)(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2147022
View Documents
Thermo Scientific Chemicals
B2147022
100 g
Each for $1,023.88
Only null left
 
Description

Description

It is used in the generation of 1,2-di(lithiomethyl)benzene. It is found to accept electrons from Li metal to give a radical anion which is highly effective in the conversion of alkyl halides to alkyllithiums. 4,4?-Di-tert-butylbiphenyl is used in production of homoallylic amine derivatives. It is also used in the preparation of lithium di-tert-butylbiphenylide, a radical anion, superior to sodium or lithium naphthalenides for metalation reactions. Along with lithium, 4,4?-Di-tert-butylbiphenyl catalyzes; reaction of chloromethyl ethyl ether and different carbonyl compounds to yield corresponding hydroxyethers and reductive opening of N-phenylazetidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the generation of 1,2-di(lithiomethyl)benzene. It is found to accept electrons from Li metal to give a radical anion which is highly effective in the conversion of alkyl halides to alkyllithiums. 4,4′-Di-tert-butylbiphenyl is used in production of homoallylic amine derivatives. It is also used in the preparation of lithium di-tert-butylbiphenylide, a radical anion, superior to sodium or lithium naphthalenides for metalation reactions. Along with lithium, 4,4′-Di-tert-butylbiphenyl catalyzes; reaction of chloromethyl ethyl ether and different carbonyl compounds to yield corresponding hydroxyethers and reductive opening of N-phenylazetidine.

Solubility
Solubility in toluene; almost transparency. Soluble in dioxane: 0.1 g/mL, (clear).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

1625-91-8
266.428
CDKCEZNPAYWORX-UHFFFAOYSA-N
74195
CC(C)(C)C1=CC=C(C=C1)C2=CC=C(C=C2)C(C)(C)C
C20H26
MFCD00008834
4,4'-di-tert-butylbiphenyl, 4,4'-di-tert-butyl-1,1'-biphenyl, 4,4'-di-t-butylbiphenyl, 1-tert-butyl-4-4-tert-butylphenyl benzene, 1,1'-biphenyl, 4,4'-bis 1,1-dimethylethyl, 1-tert-butyl-4-4-tert-butyl phenyl benzene, di-t-butylbiphenyl, pubchem9048, acmc-209st2, 4,4'-di-tertbutylbiphenyl
1-tert-butyl-4-(4-tert-butylphenyl)benzene

Specifications

1625-91-8
190°C to 192°C (13 mmHg)
MFCD00008834
2095855
CDKCEZNPAYWORX-UHFFFAOYSA-N
1-tert-butyl-4-(4-tert-butylphenyl)benzene
74195
99%
126°C to 129°C
C20H26
100 g
4,4'-di-tert-butylbiphenyl, 4,4'-di-tert-butyl-1,1'-biphenyl, 4,4'-di-t-butylbiphenyl, 1-tert-butyl-4-4-tert-butylphenyl benzene, 1,1'-biphenyl, 4,4'-bis 1,1-dimethylethyl, 1-tert-butyl-4-4-tert-butyl phenyl benzene, di-t-butylbiphenyl, pubchem9048, acmc-209st2, 4,4'-di-tertbutylbiphenyl
CC(C)(C)C1=CC=C(C=C1)C2=CC=C(C=C2)C(C)(C)C
266.428
266.43
4,4'-Di-tert-butylbiphenyl
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 216-615-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only