missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Di-n-nonyl-2,2'-bipyridine, 97%

Catalog No. AAH3147603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3147603 1 g
AAH3147606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3147603 Supplier Thermo Scientific Chemicals Supplier No. H3147603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 142646-58-0 | C28H44N2 | 408.674 g/mol

Used in the preparation of an artificial muscle with lamellar structure based on a nematic triblock copolymer. Also use as complex in cyclic voltammetric studies of copper complexes catalyzing atom transfer radical polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of an artificial muscle with lamellar structure based on a nematic triblock copolymer. Also use as complex in cyclic voltammetric studies of copper complexes catalyzing atom transfer radical polymerization.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 142646-58-0
Molecular Formula C28H44N2
Molecular Weight (g/mol) 408.674
MDL Number MFCD00800915
InChI Key VHJFWJXYEWHCGD-UHFFFAOYSA-N
Synonym 4,4'-dinonyl-2,2'-bipyridine, 4,4'-dinonyl-2,2'-bipyridyl, 4-nonyl-2-4-nonylpyridin-2-yl pyridine, 2,2'-bipyridine, 4,4'-dinonyl, 4,4'-bis nonyl-2,2'-bipyridine, dnbpy, acmc-1cctu, bpdn01, 4,4-dinonyl-2,2-bipyridine, 4,4-dinoyl-2,2-dipyridyl
PubChem CID 4285174
IUPAC Name 4-nonyl-2-(4-nonylpyridin-2-yl)pyridine
SMILES CCCCCCCCCC1=CC(=NC=C1)C2=NC=CC(=C2)CCCCCCCCC

Specifications

Melting Point 61°C to 63°C
Quantity 1 g
Formula Weight 408.67
Percent Purity 97%
Chemical Name or Material 4,4'-Di-n-nonyl-2,2'-bipyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only