missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Di(4-pyridyl)biphenyl 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 308°C |
| Color | Yellow |
| Quantity | 200 mg |
| Formula Weight | 308.38 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4,4′-Di(4-pyridyl)biphenyl |
Safety and Handling
| TSCA | No |