missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Di(1,2,3-triazolyl) Disulfide Hydrate 98.0+%, TCI America™

Catalog No. D38551G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38551G 1 g
D38555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38551G Supplier TCI America Supplier No. D38551G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6440-09-1
Molecular Formula C4H4N6S2
Molecular Weight (g/mol) 200.24
InChI Key NLSOBKJRPPRVSB-UHFFFAOYSA-N
Synonym 4,4′C-Dithiobis(1,2,3-triazole)
PubChem CID 23002801
IUPAC Name 4-(2H-triazol-4-yldisulfanyl)-2H-triazole
SMILES C1=NNN=C1SSC2=NNN=C2

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 200.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Di(1,2,3-triazolyl) Disulfide Hydrate