accessibility menu, dialog, popup

UserName

NHNNSSNHNN

CAS RN 6440-09-1

IUPAC Name: 4-(2H-1,2,3-triazol-4-yldisulfanyl)-2H-1,2,3-triazole
Synonyms: 4-(2H-1,2,3-triazol-4-yldisulfanyl)-2H-1,2,3-triazole
Molecular Weight (g/mol): 200.24
Molecular Formula: C4H4N6S2
InChi Key: NLSOBKJRPPRVSB-UHFFFAOYSA-N
SMILES: N1N=CC(SSC2=NNN=C2)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results