missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Chlorobenzoyl)pyridine, 99+%

Catalog No. AAB2255103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2255103 1 g
AAB2255106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2255103 Supplier Thermo Scientific Chemicals Supplier No. B2255103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 14548-48-2 | C12H8ClNO | 217.652 g/mol

4-(4-Chlorobenzoyl)pyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Chlorobenzoyl)pyridine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 14548-48-2
Molecular Formula C12H8ClNO
Molecular Weight (g/mol) 217.652
MDL Number MFCD00006431
InChI Key YMTFKKLFLLAFNI-UHFFFAOYSA-N
Synonym 4-4-chlorobenzoyl pyridine, 4-chlorophenyl pyridin-4-yl methanone, 4-4'-chlorobenzoyl pyridine, 4-chlorophenyl-pyridin-4-ylmethanone, methanone, 4-chlorophenyl-4-pyridinyl, acmc-1bu0c, 4-4-chloro-benzoyl pyridine, 4-chlorophenyl 4-pyridyl ketone, 4-chlorophenyl pyridin-4-ylmethanone, 4-chlorophenyl-4-pyridyl methanone
PubChem CID 98503
IUPAC Name (4-chlorophenyl)-pyridin-4-ylmethanone
SMILES C1=CC(=CC=C1C(=O)C2=CC=NC=C2)Cl

Specifications

Melting Point 107°C to 109°C
Quantity 1 g
Formula Weight 217.66
Percent Purity ≥99%
Chemical Name or Material 4-(4-Chlorobenzoyl)pyridine
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 238-587-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only