accessibility menu, dialog, popup

UserName

ClON

CAS RN 14548-48-2

IUPAC Name: 4-(4-chlorobenzoyl)pyridine
Synonyms: 4-(4-chlorobenzoyl)pyridine
Molecular Weight (g/mol): 217.65
Molecular Formula: C12H8ClNO
InChi Key: YMTFKKLFLLAFNI-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C(=O)C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results