missing translation for 'onlineSavingsMsg'
Learn More

4-[4-Chloro-3-(trifluoromethyl)phenyl]-4-hydroxypiperidine 98.0+%, TCI America™

Catalog No. C29095G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29095G 5 g
C290925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29095G Supplier TCI America Supplier No. C29095G

Chemical Identifiers

CAS 21928-50-7
Molecular Formula C12H13ClF3NO
Molecular Weight (g/mol) 279.687
MDL Number MFCD00066999
InChI Key RALRVIPTUXSBPO-UHFFFAOYSA-N
Synonym 4-4-chloro-3-trifluoromethyl phenyl-4-piperidinol, 4-4-chloro-3-trifluoromethyl phenyl piperidin-4-ol, 4-piperidinol, 4-4-chloro-3-trifluoromethyl phenyl, 4-4-chloro-3-trifluoromethyl phenyl-4-hydroxypiperidine, piperidine alcohol, 4-4-chloro-3-trifluoromethylphenyl-4-piperidinol, piperidinalkohol
PubChem CID 89107
IUPAC Name 4-[4-chloro-3-(trifluoromethyl)phenyl]piperidin-4-ol
SMILES C1CNCCC1(C2=CC(=C(C=C2)Cl)C(F)(F)F)O

Specifications

Melting Point 140°C
Color Yellow
Quantity 5 g
Formula Weight 279.69
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-[4-Chloro-3-(trifluoromethyl)phenyl]-4-hydroxypiperidine