missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Butylphenylazo)phenol 98.0+%, TCI America™

Catalog No. B23575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23575G 5 g
B235725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23575G Supplier TCI America Supplier No. B23575G
Only null left

Chemical Identifiers

CAS 2496-21-1
Molecular Formula C16H18N2O
Molecular Weight (g/mol) 254.333
MDL Number MFCD00430795
InChI Key MHSQAHVLQVTXKH-UHFFFAOYSA-N
PubChem CID 5705751
IUPAC Name 4-[(4-butylphenyl)hydrazinylidene]cyclohexa-2,5-dien-1-one
SMILES CCCCC1=CC=C(C=C1)NN=C2C=CC(=O)C=C2

Specifications

Melting Point 80°C
Quantity 5 g
Synonym 4-Butyl-4′C-hydroxyazobenzene
Formula Weight 254.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Butylphenylazo)phenol