missing translation for 'onlineSavingsMsg'
Learn More

4-[(4-bromothien-2-yl)methyl]morpholine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 194851-19-9
Molecular Formula C9H12BrNOS
Molecular Weight (g/mol) 262.17
MDL Number MFCD01653482
InChI Key XYQVGDMXJNORKK-UHFFFAOYSA-N
Synonym 4-4-bromothiophen-2-yl methyl morpholine, 4-4-bromothien-2-yl methyl morpholine, 4-bromo-2-morpholin-4-ylmethyl thiophene, cambridge id 5430345, 4-bromo-2-morpholinomethylthiophene, 4-4-bromo-thiophen-2-ylmethyl-morpholine, morpholine, 4-4-bromo-2-thienyl methyl
PubChem CID 783497
IUPAC Name 4-[(4-bromothiophen-2-yl)methyl]morpholine
SMILES BrC1=CSC(CN2CCOCC2)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC74210DA
SDS View Documents
Thermo Scientific Chemicals
CC74210DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC74210DE
SDS View Documents
Thermo Scientific Chemicals
CC74210DE
5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

194851-19-9
262.17
XYQVGDMXJNORKK-UHFFFAOYSA-N
783497
BrC1=CSC(CN2CCOCC2)=C1
C9H12BrNOS
MFCD01653482
4-4-bromothiophen-2-yl methyl morpholine, 4-4-bromothien-2-yl methyl morpholine, 4-bromo-2-morpholin-4-ylmethyl thiophene, cambridge id 5430345, 4-bromo-2-morpholinomethylthiophene, 4-4-bromo-thiophen-2-ylmethyl-morpholine, morpholine, 4-4-bromo-2-thienyl methyl
4-[(4-bromothiophen-2-yl)methyl]morpholine

Specifications

194851-19-9
100.0
Amber Glass Bottle
MFCD01653482
4-4-bromothiophen-2-yl methyl morpholine, 4-4-bromothien-2-yl methyl morpholine, 4-bromo-2-morpholin-4-ylmethyl thiophene, cambridge id 5430345, 4-bromo-2-morpholinomethylthiophene, 4-4-bromo-thiophen-2-ylmethyl-morpholine, morpholine, 4-4-bromo-2-thienyl methyl
BrC1=CSC(CN2CCOCC2)=C1
262.17
262.17
4-[(4-Bromothien-2-yl)methyl]morpholine
96.5
97%
C9H12BrNOS
1g
XYQVGDMXJNORKK-UHFFFAOYSA-N
4-[(4-bromothiophen-2-yl)methyl]morpholine
783497
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'