Learn More
4-[(4-bromothien-2-yl)methyl]morpholine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC74210DA
Specifications
| 4-[(4-Bromothien-2-yl)methyl]morpholine | |
| 96.5 | |
| 97% | |
| C9H12BrNOS | |
| 1g | |
| XYQVGDMXJNORKK-UHFFFAOYSA-N | |
| 4-[(4-bromothiophen-2-yl)methyl]morpholine | |
| 783497 | |
| 97% |
| 194851-19-9 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD01653482 | |
| 4-4-bromothiophen-2-yl methyl morpholine, 4-4-bromothien-2-yl methyl morpholine, 4-bromo-2-morpholin-4-ylmethyl thiophene, cambridge id 5430345, 4-bromo-2-morpholinomethylthiophene, 4-4-bromo-thiophen-2-ylmethyl-morpholine, morpholine, 4-4-bromo-2-thienyl methyl | |
| BrC1=CSC(CN2CCOCC2)=C1 | |
| 262.17 | |
| 262.17 |
Chemical Identifiers
| 194851-19-9 | |
| 262.17 | |
| XYQVGDMXJNORKK-UHFFFAOYSA-N | |
| 783497 | |
| BrC1=CSC(CN2CCOCC2)=C1 |
| C9H12BrNOS | |
| MFCD01653482 | |
| 4-4-bromothiophen-2-yl methyl morpholine, 4-4-bromothien-2-yl methyl morpholine, 4-bromo-2-morpholin-4-ylmethyl thiophene, cambridge id 5430345, 4-bromo-2-morpholinomethylthiophene, 4-4-bromo-thiophen-2-ylmethyl-morpholine, morpholine, 4-4-bromo-2-thienyl methyl | |
| 4-[(4-bromothiophen-2-yl)methyl]morpholine |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger