missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Bromophenylazo)phenol 98.0+%, TCI America™

Catalog No. B4053200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4053200MG 200 mg
B40531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4053200MG Supplier TCI America Supplier No. B4053200MG
Only null left

Chemical Identifiers

CAS 3035-94-7
Molecular Formula C12H9BrN2O
Molecular Weight (g/mol) 277.121
MDL Number MFCD00093929
InChI Key BGGPPMHBCFPATP-UHFFFAOYSA-N
Synonym 4-Bromo-4′C-hydroxyazobenzene
PubChem CID 5492867
IUPAC Name 4-[(4-bromophenyl)hydrazinylidene]cyclohexa-2,5-dien-1-one
SMILES C1=CC(=O)C=CC1=NNC2=CC=C(C=C2)Br

Specifications

Melting Point 161°C
Quantity 200 mg
Formula Weight 277.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Bromophenylazo)phenol
RTECSNumber SJ8380000
TSCA No