missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Bromophenyl)morpholine 98.0+%, TCI America™
$223.33 - $223.33
Chemical Identifiers
| CAS | 30483-75-1 |
|---|---|
| Molecular Formula | C10H12BrNO |
| Molecular Weight (g/mol) | 242.116 |
| MDL Number | MFCD04112483 |
| InChI Key | UJTKZWNRUPTHSB-UHFFFAOYSA-N |
| PubChem CID | 11622851 |
| IUPAC Name | 4-(4-bromophenyl)morpholine |
| SMILES | C1COCCN1C2=CC=C(C=C2)Br |
Chemical Identifiers
| 30483-75-1 | |
| 242.116 | |
| UJTKZWNRUPTHSB-UHFFFAOYSA-N | |
| 4-(4-bromophenyl)morpholine |
| C10H12BrNO | |
| MFCD04112483 | |
| 11622851 | |
| C1COCCN1C2=CC=C(C=C2)Br |
Specifications
| 30483-75-1 | |
| C10H12BrNO | |
| 5 g | |
| C1COCCN1C2=CC=C(C=C2)Br | |
| 242.116 | |
| 242.12 | |
| Crystalline Powder |
| 115°C | |
| MFCD04112483 | |
| UJTKZWNRUPTHSB-UHFFFAOYSA-N | |
| 4-(4-bromophenyl)morpholine | |
| 11622851 | |
| ≥98.0% (GC,T) | |
| 4-(4-Bromophenyl)morpholine |
Safety and Handling
TSCA : No