missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Bromophenyl)dibenzothiophene 98.0+%, TCI America™

Catalog No. p-7124062
Change view
Click to view available options
Quantity:
200 mg
1 g
Catalog No. Quantity
B5384200MG 200 mg
B53841G 1 g
2 options

Catalog No. B5384200MG

Supplier: TCI America B5384200MG

Only null left

Chemical Identifiers

CAS 530402-77-8
Molecular Formula C18H11BrS
Molecular Weight (g/mol) 339.25
MDL Number MFCD19441307
InChI Key FIAXBPCUAUSGJH-UHFFFAOYSA-N
PubChem CID 59866463
IUPAC Name 4-(4-bromophenyl)dibenzothiophene
SMILES C1=CC=C2C(=C1)C3=C(S2)C(=CC=C3)C4=CC=C(C=C4)Br

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 200 mg
Formula Weight 339.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Bromophenyl)dibenzothiophene