missing translation for 'onlineSavingsMsg'
Learn More

4'-(4-Bromophenyl)-2,2':6',2″-terpyridine 97.0+%, TCI America™

Catalog No. B32181G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32181G 1 g
B32185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32181G Supplier TCI America Supplier No. B32181G

Chemical Identifiers

CAS 89972-76-9
Molecular Formula C21H14BrN3
Molecular Weight (g/mol) 388.268
MDL Number MFCD06796984
InChI Key BOPPHKMZOYPASP-UHFFFAOYSA-N
Synonym 4'-4-bromophenyl-2,2':6',2-terpyridine, 4'-4-bromophenyl-2,6':2',2-terpyridine, 4'-4-bromophenyl-alpha,alpha',alpha-tripyridyl, 4-4-bromophenyl-6-pyridin-2-yl-2,2'-bipyridine, acmc-209r3y, 4'-4-bromophenyl-2,2':6',2'-terpyridine, 4-4'-bromophenyl-2,6-di 2-pyridyl pyridine, 4-4-bromophenyl-2,6-bis 2-pyridyl pyridine
PubChem CID 10762837
IUPAC Name 4-(4-bromophenyl)-2,6-dipyridin-2-ylpyridine
SMILES C1=CC=NC(=C1)C2=CC(=CC(=N2)C3=CC=CC=N3)C4=CC=C(C=C4)Br

Specifications

Melting Point 162°C
Color White-Yellow
Quantity 1 g
Formula Weight 388.27
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-(4-Bromophenyl)-2,2′:6′,2′′-terpyridine