accessibility menu, dialog, popup

UserName

NNNBr

CAS RN 89972-76-9

IUPAC Name: 4-(4-bromophenyl)-6-(pyridin-2-yl)-2,2'-bipyridine
Synonyms: 4-(4-bromophenyl)-6-(pyridin-2-yl)-2,2'-bipyridine
Molecular Weight (g/mol): 388.27
Molecular Formula: C21H14BrN3
InChi Key: BOPPHKMZOYPASP-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)