missing translation for 'onlineSavingsMsg'
Learn More

4'-(4-Bromophenyl)-2,2':6',2″-terpyridine, 97%

Catalog No. AAH3777803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3777803 1 g
AAH3777806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3777803 Supplier Thermo Scientific Chemicals Supplier No. H3777803

CAS: 89972-76-9 | C21H14BrN3 | 388.268 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89972-76-9
Molecular Formula C21H14BrN3
Molecular Weight (g/mol) 388.268
MDL Number MFCD06796984
InChI Key BOPPHKMZOYPASP-UHFFFAOYSA-N
Synonym 4'-4-bromophenyl-2,2':6',2-terpyridine, 4'-4-bromophenyl-2,6':2',2-terpyridine, 4'-4-bromophenyl-alpha,alpha',alpha-tripyridyl, 4-4-bromophenyl-6-pyridin-2-yl-2,2'-bipyridine, acmc-209r3y, 4'-4-bromophenyl-2,2':6',2'-terpyridine, 4-4'-bromophenyl-2,6-di 2-pyridyl pyridine, 4-4-bromophenyl-2,6-bis 2-pyridyl pyridine
PubChem CID 10762837
IUPAC Name 4-(4-bromophenyl)-2,6-dipyridin-2-ylpyridine
SMILES C1=CC=NC(=C1)C2=CC(=CC(=N2)C3=CC=CC=N3)C4=CC=C(C=C4)Br

Specifications

Melting Point 143°C to 145°C
Quantity 1 g
Formula Weight 388.27
Percent Purity 98%
Chemical Name or Material 4'-(4-Bromophenyl)-2,6':2',2''-terpyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 679-534-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only