missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Bromophenyl)-1,2,3-thiadiazole, 97%, Thermo Scientific™

$65.89 - $65.89

Chemical Identifiers

CAS 40753-13-7
Molecular Formula C8H5BrN2S
Molecular Weight (g/mol) 241.106
MDL Number MFCD00084907
InChI Key HGWOTVRPRHVJQK-UHFFFAOYSA-N
PubChem CID 218520
IUPAC Name 4-(4-bromophenyl)thiadiazole
SMILES C1=CC(=CC=C1C2=CSN=N2)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC16410DA
SDS View Documents
Thermo Scientific Chemicals
CC16410DA
1 g Amber Glass Bottle
Each for $65.89
Only null left
 
Description

Description

Specifications

Chemical Identifiers

40753-13-7
241.106
HGWOTVRPRHVJQK-UHFFFAOYSA-N
4-(4-bromophenyl)thiadiazole
C8H5BrN2S
MFCD00084907
218520
C1=CC(=CC=C1C2=CSN=N2)Br

Specifications

40753-13-7
100.0
Amber Glass Bottle
MFCD00084907
4-4-bromophenyl-1,2,3-thiadiazole, 1,2,3-thiadiazole, 4-4-bromophenyl, 1,2,3-thiadiazole,4-4-bromophenyl, 4-4-bromophenyl thiadiazole
C1=CC(=CC=C1C2=CSN=N2)Br
241.106
241.11
4-(4-Bromophenyl)-1, 2, 3-thiadiazole
96.5
97%
C8H5BrN2S
1 g
HGWOTVRPRHVJQK-UHFFFAOYSA-N
4-(4-bromophenyl)thiadiazole
218520
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'