missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Bromophényl)-1,2,3-thiadiazole, 97%, Thermo Scientific™

Numéro de catalogue. CC16410DA
Modifier l'affichage
Click to view available options
Quantité:
1 g
Emballage:
Bouteille en verre ambrée
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC16410DA 1 g Bouteille en verre ambrée
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC16410DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC16410DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 40753-13-7
Formule moléculaire C8H5BrN2S
Poids moléculaire (g/mol) 241.106
Numéro MDL MFCD00084907
Clé InChI HGWOTVRPRHVJQK-UHFFFAOYSA-N
PubChem CID 218520
Nom de l’IUPAC 4-(4-bromophényl)thiadiazole
SOURIRES C1=CC(=CC=C1C2=CSN=N2)Br

Spécifications

Nom ou substance chimique 4-(4-Bromophenyl)-1, 2, 3-thiadiazole
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Bouteille en verre ambrée
Quantité 1 g
Synonyme 4-4-bromophenyl-1,2,3-thiadiazole, 1,2,3-thiadiazole, 4-4-bromophenyl, 1,2,3-thiadiazole,4-4-bromophenyl, 4-4-bromophenyl thiadiazole
Poids de formule 241.11
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.