missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bismaleimidodiphenylmethane 96.0+%, TCI America™

Catalog No. B110925G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B110925G 25 g
B1109100G 100 g
B1109500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B110925G Supplier TCI America Supplier No. B110925G

Chemical Identifiers

CAS 13676-54-5
Molecular Formula C21H14N2O4
Molecular Weight (g/mol) 358.353
MDL Number MFCD00005507
InChI Key XQUPVDVFXZDTLT-UHFFFAOYSA-N
Synonym bismaleimide, bismaleimide s, bis 4-maleimidophenyl methane, diphenylmethanebismaleimide, p,p'-dimaleimidodiphenylmethane, 4,4'-methylenebis n-phenylmaleimide, bis p-maleimidophenyl methane, 4,4'-dimaleimidodiphenylmethane, 4,4'-diphenylmethanedimaleimide, 4,4'-biphenylmethanebismaleimide
PubChem CID 83648
IUPAC Name 1-[4-[[4-(2,5-dioxopyrrol-1-yl)phenyl]methyl]phenyl]pyrrole-2,5-dione
SMILES C1=CC(=CC=C1CC2=CC=C(C=C2)N3C(=O)C=CC3=O)N4C(=O)C=CC4=O

Specifications

Melting Point 159°C
Color Yellow
Quantity 25 g
UN Number 2811
Formula Weight 358.35
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bismaleimidodiphenylmethane
EINECSNumber (5)-3377
RTECSNumber ON5670000
TSCA Yes