missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(hydroxymethyl)-2,2'-bipyridine 95.0+%, TCI America™

Catalog No. B35115G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35115G 5 g
B35111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35115G Supplier TCI America Supplier No. B35115G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109073-77-0
Molecular Formula C12H12N2O2
Molecular Weight (g/mol) 216.24
MDL Number MFCD06202808
InChI Key RRXGRDMHWYLJSY-UHFFFAOYSA-N
PubChem CID 11160239
IUPAC Name [2-[4-(hydroxymethyl)pyridin-2-yl]pyridin-4-yl]methanol
SMILES C1=CN=C(C=C1CO)C2=NC=CC(=C2)CO

Specifications

Melting Point 174°C
Color White
Quantity 5 g
Synonym 2,2′-Bipyridine-4,4′-dimethanol, 4,4′-Bis(hydroxymethyl)-2,2′-bipyridyl
Formula Weight 216.24
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(hydroxymethyl)-2,2′-bipyridine