accessibility menu, dialog, popup

UserName

CAS RN 109073-77-0

IUPAC Name: [4'-(hydroxymethyl)-[2,2'-bipyridin]-4-yl]methanol
Synonyms: [4'-(hydroxymethyl)-[2,2'-bipyridin]-4-yl]methanol
Molecular Weight (g/mol): 216.24
Molecular Formula: C12H12N2O2
InChi Key: RRXGRDMHWYLJSY-UHFFFAOYSA-N
SMILES: OCC1=CC(=NC=C1)C1=NC=CC(CO)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results