missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(diethylamino)benzophenone 98.0+%, TCI America™

Catalog No. B0139500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0139500G 500 g
B013925G 25 g
B0139100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0139500G Supplier TCI America Supplier No. B0139500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 90-93-7
Molecular Formula C21H28N2O
Molecular Weight (g/mol) 324.47
MDL Number MFCD00009044
InChI Key VYHBFRJRBHMIQZ-UHFFFAOYSA-N
Synonym 4,4'-bis diethylamino benzophenone, michler's ethyl ketone, bis 4-diethylamino phenyl methanone, methanone, bis 4-diethylamino phenyl, 4,4'-tetraethyldiamino benzophenone, p,p'-bis diethylamino benzophenone, p,p'-tetraethyldiamino benzophenone, ethyl michler ketone
PubChem CID 66663
IUPAC Name 4-[4-(diethylamino)benzoyl]-N,N-diethylaniline
SMILES CCN(CC)C1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)N(CC)CC

Specifications

Melting Point 94°C
Quantity 500 g
UN Number 3077
Formula Weight 324.47
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(diethylamino)benzophenone
EINECSNumber (4)-0893
RTECSNumber OS9545500
TSCA Yes