missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 4,4'-Bis(diethylamino)benzophenone

Catalog No. 160326100G
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
160326100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 160326100G Supplier Sigma Aldrich Supplier No. 160326100G
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4,4'-Bis(diethylamino)benzophenone

Specifications

CAS 90-93-7
Melting Point 89°C to 92°C
Molecular Formula C21H28N2O
Linear Formula [(C2H5)2 NC6H4]2 CO
MDL Number MFCD00009044
Quantity 100 g
Synonym 4,4 -(Tetraethyldiamino)benzophenone; 4,4 -Bis(diethylamino)diphenyl ketone; p ,p -Tetraethyldiaminobenzophenone; Bis[4-(diethylamino)phenyl]methanone; Michlers ethyl ketone
Molecular Weight (g/mol) 324.46
Percent Purity ≥99%