missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(bromomethyl)biphenyl 95.0+%, TCI America™

Catalog No. p-7119356
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19211G 1 g
B19215G 5 g
B192125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B19211G Supplier TCI America Supplier No. B19211G
Only null left

Chemical Identifiers

CAS 20248-86-6
Molecular Formula C14H12Br2
Molecular Weight (g/mol) 340.06
MDL Number MFCD00094419
InChI Key HMUGRILXVBKBID-UHFFFAOYSA-N
PubChem CID 621238
IUPAC Name 4,4'-bis(bromomethyl)-1,1'-biphenyl
SMILES BrCC1=CC=C(C=C1)C1=CC=C(CBr)C=C1

Specifications

Melting Point 171°C
Quantity 1 g
UN Number 1759
Formula Weight 340.06
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(bromomethyl)biphenyl