missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Bis(alpha,alpha-dimethylbenzyl)diphenylamine 98.0+%, TCI America™
Supplier: TCI America B1970500G
Specifications
| 4,4′-Bis(alpha,alpha-dimethylbenzyl)diphenylamine | |
| 100°C | |
| C30H31N | |
| 500 g | |
| CC(C)(C1=CC=CC=C1)C2=CC=C(C=C2)NC3=CC=C(C=C3)C(C)(C)C4=CC=CC=C4 | |
| 405.585 | |
| 405.59 | |
| Crystalline Powder |
| 10081-67-1 | |
| White-Yellow | |
| MFCD00337918 | |
| UJAWGGOCYUPCPS-UHFFFAOYSA-N | |
| 4-(2-phenylpropan-2-yl)-N-[4-(2-phenylpropan-2-yl)phenyl]aniline | |
| 82343 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 10081-67-1 | |
| 405.585 | |
| UJAWGGOCYUPCPS-UHFFFAOYSA-N | |
| 4-(2-phenylpropan-2-yl)-N-[4-(2-phenylpropan-2-yl)phenyl]aniline |
| C30H31N | |
| MFCD00337918 | |
| 82343 | |
| CC(C)(C1=CC=CC=C1)C2=CC=C(C=C2)NC3=CC=C(C=C3)C(C)(C)C4=CC=CC=C4 |
Safety and Handling
EINECSNumber : (4)-0238
TSCA : Yes