missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(alpha,alpha-dimethylbenzyl)diphenylamine 98.0+%, TCI America™

Catalog No. B197025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B197025G 25 g
B1970500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B197025G Supplier TCI America Supplier No. B197025G
Only null left

Chemical Identifiers

CAS 10081-67-1
Molecular Formula C30H31N
Molecular Weight (g/mol) 405.585
MDL Number MFCD00337918
InChI Key UJAWGGOCYUPCPS-UHFFFAOYSA-N
PubChem CID 82343
IUPAC Name 4-(2-phenylpropan-2-yl)-N-[4-(2-phenylpropan-2-yl)phenyl]aniline
SMILES CC(C)(C1=CC=CC=C1)C2=CC=C(C=C2)NC3=CC=C(C=C3)C(C)(C)C4=CC=CC=C4

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 25 g
Formula Weight 405.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(alpha,alpha-dimethylbenzyl)diphenylamine
EINECSNumber (4)-0238
TSCA Yes