missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Bis(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)biphenyl 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 248°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 378.08 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4,4′-Bis(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)biphenyl |
Safety and Handling
| TSCA | No |