accessibility menu, dialog, popup

UserName

CAS RN 5487-93-4

BOOH3CCH3BOOCH3H3C

CAS RN 5487-93-4

IUPAC Name: 2-[4'-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-[1,1'-biphenyl]-4-yl]-5,5-dimethyl-1,3,2-dioxaborinane
Synonyms: 2-[4'-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-[1,1'-biphenyl]-4-yl]-5,5-dimethyl-1,3,2-dioxaborinane
Molecular Weight (g/mol): 378.08
Molecular Formula: C22H28B2O4
InChi Key: NJXUWONHNYJPMY-UHFFFAOYSA-N
SMILES: CC1(C)COB(OC1)C1=CC=C(C=C1)C1=CC=C(C=C1)B1OCC(C)(C)CO1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results