missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(2-methoxystyryl)biphenyl 90.0+%, TCI America™

Catalog No. B273025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B273025G 25 g
B2730500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B273025G Supplier TCI America Supplier No. B273025G
Only null left

Chemical Identifiers

CAS 40470-68-6
Molecular Formula C30H26O2
Molecular Weight (g/mol) 418.536
InChI Key HZAWHDJKNZWAAR-BMJUYKDLSA-N
PubChem CID 86307593
IUPAC Name 1-methoxy-2-[(Z)-2-[4-[4-[(Z)-2-(2-methoxyphenyl)ethenyl]phenyl]phenyl]ethenyl]benzene
SMILES COC1=CC=CC=C1C=CC2=CC=C(C=C2)C3=CC=C(C=C3)C=CC4=CC=CC=C4OC

Specifications

Melting Point 218°C
Quantity 25 g
Formula Weight 418.54
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(2-methoxystyryl)biphenyl