missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4-Bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b']dithiophene 98.0+%, TCI America™
$915.25 - $3180.08
Chemical Identifiers
| CAS | 365547-20-2 |
|---|---|
| Molecular Formula | C25H38S2 |
| Molecular Weight (g/mol) | 402.699 |
| InChI Key | NUCIQEWGTLOQTR-UHFFFAOYSA-N |
| PubChem CID | 59426893 |
| SMILES | CCCCC(CC)CC1(C2=C(C3=C1C=CS3)SC=C2)CC(CC)CCCC |
Chemical Identifiers
| 365547-20-2 | |
| 402.699 | |
| 59426893 |
| C25H38S2 | |
| NUCIQEWGTLOQTR-UHFFFAOYSA-N | |
| CCCCC(CC)CC1(C2=C(C3=C1C=CS3)SC=C2)CC(CC)CCCC |
Specifications
| 365547-20-2 | |
| 1 g | |
| NUCIQEWGTLOQTR-UHFFFAOYSA-N | |
| 402.699 | |
| 402.70 | |
| Liquid |
| C25H38S2 | |
| 4,4-Bis(2-ethylhexyl)-4H-cyclopenta[1,2-b:5,4-b′C]dithiophene | |
| CCCCC(CC)CC1(C2=C(C3=C1C=CS3)SC=C2)CC(CC)CCCC | |
| 59426893 | |
| ≥98.0% (GC) | |
| 4,4-Bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b′]dithiophene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator