accessibility menu, dialog, popup

UserName

CAS RN 365547-20-2

IUPAC Name: 7,7-bis(2-ethylhexyl)-3,11-dithiatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraene
Synonyms: 7,7-bis(2-ethylhexyl)-3,11-dithiatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraene
Molecular Weight (g/mol): 402.7
Molecular Formula: C25H38S2
InChi Key: NUCIQEWGTLOQTR-UHFFFAOYNA-N
SMILES: CCCCC(CC)CC1(CC(CC)CCCC)C2=C(SC=C2)C2=C1C=CS2
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results